Search
Search

Mrto4| Gene Symbol: Mrto4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290810.1 | NP_001277739.1 | 3 - 4 | 474 | 62 | |
| NM_023536.3 | NP_076025.1 | 3 - 4 | 474 | 62 | ||
| GenBank mRNA | AK011387.1 | - | 3 - 4 | 196 | 62 | |
| BC005734.1 | - | 3 - 4 | 196 | 3598883 | 62 | |
| BC096556.1 | - | 3 - 4 | 218 | 62 | ||
| BC138221.1 | - | 3 - 4 | 273 | 8861240 | 62 | |
| BC145205.1 | - | 3 - 4 | 273 | 9053756 | 62 | |
| BC145209.1 | - | 3 - 4 | 295 | 62 | ||
Set Membership: |
|