Search
Search

Txndc15| Gene Symbol: Txndc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175150.3 | NP_780359.2 | 3 - 4 | 835 | 62 | |
| GenBank mRNA | AK079517.1 | - | 3 - 4 | 782 | 62 | |
| AK160051.1 | - | 3 - 4 | 804 | 62 | ||
| AK167701.1 | - | 3 - 4 | 743 | 62 | ||
| AK167838.1 | - | 3 - 4 | 801 | 62 | ||
| AK167977.1 | - | 3 - 4 | 800 | 62 | ||
| BC029015.1 | - | 2 - 3 | 571 | 62 | ||
| BC038082.1 | - | 1 - 2 | 93 | 62 | ||
| BC062180.1 | - | 3 - 4 | 835 | 6336176 | 62 | |
| BC094275.1 | - | 3 - 4 | 671 | 62 | ||
Set Membership: |
|