Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01342200_m1
          See other TMEM207 GE Assays ›
          Gene Symbol:
          Tmem207
          Gene Name:
          transmembrane protein 207
          Gene Aliases:
          Gm540
          Chromosome Location:
          Chr.16: 26472713 - 26526839 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01342200_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem207
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101640.1 NP_001095110.1 4 - 5 360 92
          GenBank mRNA AK142524.1 - 3 - 4 1376 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001421.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P340628 NM_001101640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P340628 AK142524.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P340628 NM_001101640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P340628 AK142524.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826640 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 156574 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124836 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P340628 NM_001101640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P340628 AK142524.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P340628 NM_001101640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P340628 AK142524.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08541076 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124836 - -
          Whole Mouse Genome G4122A A_52_P340628 NM_001101640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P340628 AK142524.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P340628 NM_001101640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P340628 AK142524.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30620995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30620993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30620994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12608635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30620990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12566266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12566259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12878819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30620992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12878825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30620996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30620999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30621110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12788168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12904265 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438779 - -
          Affymetrix HT MG-430 PM Array Plate 1445371_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10438778 - -
          Mouse Genome 430 2.0 Array 1445371_PM_at - -
          Mouse Genome 430 2.0 Array 1445371_at - -
          Affymetrix Mouse Clariom S Array PSR1600011744.mm.1 NM_001101640.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600011744.mm.1 NM_001101640.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600011744.mm.1 TC1600001421.mm.1 MTA TC
          Mouse Expression Set 430 1445371_at - -
          Mouse Expression Set 430 1445371_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445371_at - -
          Affymetrix HT Mouse Genome 430B Array 1445371_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 410414 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline