Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01342235_mH
          See other CYP4F37 GE Assays ›
          Gene Symbol:
          Cyp4f37
          Gene Name:
          cytochrome P450, family 4, subfamily f, polypeptide 37
          Gene Aliases:
          Gm9705
          Chromosome Location:
          Chr.17: 32621319 - 32636184 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01342235_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp4f37
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100187.1 NP_001093657.1 3 - 4 380 92
          GenBank mRNA AK132492.1 - 3 - 4 380 92
          BC151030.1 - 365 9087943 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000565.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Process(es)

          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143489 NM_001100187.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143489 AK132492.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 711499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74157 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4882 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143489 NM_001100187.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143489 AK132492.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08761516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31146259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13260388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13268146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13260387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31146222 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10443878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443879 - -
          Affymetrix Mouse Gene 1.1 ST Array 10443876 - -
          Affymetrix Mouse Clariom S Array PSR1700005146.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005146.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005146.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005141.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005141.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005141.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005143.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005143.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005143.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005145.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005145.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005145.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005142.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005142.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005142.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005144.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005144.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005144.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005140.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005140.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005140.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005149.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005149.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005149.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005137.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005137.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005137.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005138.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005138.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005138.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005147.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005147.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005147.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005148.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005148.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005148.mm.1 TC1700000565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005139.mm.1 NM_001100187.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005139.mm.1 NM_001100187.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005139.mm.1 TC1700000565.mm.1 MTA TC
          Mouse Expression Set 430 1430172_a_at - -
          Mouse Expression Set 430 1430172_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1430172_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1430172_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 534253 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0