| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600000028.mm.1 |
NM_148924.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000028.mm.1 |
NM_148924.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000028.mm.1 |
TC1600000009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000025.mm.1 |
NM_148924.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000025.mm.1 |
NM_148924.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000025.mm.1 |
TC1600000009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000029.mm.1 |
NM_148924.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000029.mm.1 |
NM_148924.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000029.mm.1 |
TC1600000009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000023.mm.1 |
NM_148924.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000023.mm.1 |
NM_148924.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000023.mm.1 |
TC1600000009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000026.mm.1 |
NM_148924.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000026.mm.1 |
NM_148924.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000026.mm.1 |
TC1600000009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000027.mm.1 |
NM_148924.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000027.mm.1 |
NM_148924.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000027.mm.1 |
TC1600000009.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000033.mm.1 |
NM_148924.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000033.mm.1 |
NM_148924.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000033.mm.1 |
TC1600000009.mm.1 |
MTA TC |