| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900027349.mm.1 |
NM_001205321.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027349.mm.1 |
NM_001205321.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027349.mm.1 |
NM_009134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027349.mm.1 |
NM_009134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027349.mm.1 |
TC0900003245.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900027350.mm.1 |
NM_001205321.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027350.mm.1 |
NM_001205321.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027350.mm.1 |
NM_009134.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027350.mm.1 |
NM_009134.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027350.mm.1 |
TC0900003245.mm.1 |
MTA TC |