Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01342822_m1
          See other LSG1 GE Assays ›
          Gene Symbol:
          Lsg1
          Gene Name:
          large subunit GTPase 1 homolog (S. cerevisiae)
          Gene Aliases:
          5830465I20, AA409273, D16Bwg1547e
          Chromosome Location:
          Chr.16: 30561369 - 30587589 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01342822_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178069.5 NP_835170.1 6 - 7 638 61
          GenBank mRNA AK030940.1 - 6 - 7 613 61
          AK078501.1 - 6 - 7 611 61
          AK132527.1 - 7 - 8 841 61
          AK135345.1 - 633 61
          AK146895.1 - 6 - 7 613 61
          BC043724.1 - 6 - 7 638 5004563 61
          BC080306.1 - 4 - 5 369 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001466.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lsg1

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          metabolic process
          protein transport
          ribosome biogenesis
          nuclear export
          nucleotide binding
          GTPase activity
          thiamine-pyrophosphatase activity
          GTP binding
          8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
          UDP-2,3-diacylglucosamine hydrolase activity
          bis(5'-nucleosyl)-tetraphosphatase activity
          dATP pyrophosphohydrolase activity
          pyrophosphatase activity
          hydrolase activity
          dihydroneopterin monophosphate phosphatase activity
          dihydroneopterin triphosphate pyrophosphohydrolase activity
          dITP diphosphatase activity
          dTTP diphosphatase activity
          XTP diphosphatase activity
          ATP-dependent 5'-3' DNA helicase activity
          phosphocholine hydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600012096.mm.1 NM_178069.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012096.mm.1 NM_178069.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012096.mm.1 TC1600001466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012097.mm.1 NM_178069.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012097.mm.1 NM_178069.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012097.mm.1 TC1600001466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012095.mm.1 NM_178069.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012095.mm.1 NM_178069.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012095.mm.1 TC1600001466.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline