| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800018523.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018523.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018523.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018532.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018532.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018532.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018535.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018535.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018535.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018524.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018524.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018524.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018533.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018533.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018533.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018528.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018528.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018528.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018520.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018520.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018520.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018522.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018522.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018522.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018527.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018527.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018527.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018531.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018531.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018531.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018521.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018521.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018521.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018525.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018525.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018525.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018529.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018529.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018529.mm.1 |
TC0800002428.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018534.mm.1 |
NM_001164679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018534.mm.1 |
NM_001164679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018534.mm.1 |
TC0800002428.mm.1 |
MTA TC |