Search
Search

Dppa4| Gene Symbol: Dppa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018002.1 | NP_001018002.1 | 4 - 5 | 587 | 68 | |
| NM_028610.2 | NP_082886.2 | 5 - 6 | 743 | 68 | ||
| GenBank mRNA | AF490348.1 | - | 5 - 6 | 660 | 68 | |
| AK033197.1 | - | 5 - 6 | 744 | 68 | ||
| BC064754.1 | - | 5 - 6 | 682 | 30024281 | 68 | |
| BY731814.1 | - | 4 - 5 | 553 | 68 | ||
Set Membership: |
|