Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01343391_gH
          See other DPPA2 GE Assays ›
          Gene Symbol:
          Dppa2
          Gene Name:
          developmental pluripotency associated 2
          Gene Aliases:
          2410088E07Rik, C80932, D19Mgi18, ECAT15-2
          Chromosome Location:
          Chr.16: 48310274 - 48319513 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01343391_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dppa2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028615.1 NP_082891.1 2 - 3 404 90
          GenBank mRNA AF490346.1 - 2 - 3 202 90
          AK010743.1 - 2 - 3 202 90
          AK145645.1 - 2 - 3 231 90
          AK145760.1 - 2 - 3 201 90
          AK163375.1 - 2 - 3 204 90
          AK167211.1 - 2 - 3 202 90
          AY561246.1 - 2 - 3 404 90
          BC137768.1 - 2 - 3 207 8860787 90

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dppa2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          stem cell population maintenance
          regulation of histone methylation
          lung-associated mesenchyme development
          positive regulation of stem cell proliferation
          chromatin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600006028.mm.1 NM_028615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006028.mm.1 NM_028615.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006028.mm.1 TC1600000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006032.mm.1 NM_028615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006032.mm.1 NM_028615.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006032.mm.1 TC1600000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006033.mm.1 NM_028615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006033.mm.1 NM_028615.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006033.mm.1 TC1600000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006029.mm.1 NM_028615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006029.mm.1 NM_028615.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006029.mm.1 TC1600000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006030.mm.1 NM_028615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006030.mm.1 NM_028615.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006030.mm.1 TC1600000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006031.mm.1 NM_028615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006031.mm.1 NM_028615.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006031.mm.1 TC1600000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006025.mm.1 NM_028615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006025.mm.1 NM_028615.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006025.mm.1 TC1600000693.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006027.mm.1 NM_028615.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006027.mm.1 NM_028615.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006027.mm.1 TC1600000693.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 885126 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline