Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01343539_m1
          See other FBXO40 GE Assays ›
          Gene Symbol:
          Fbxo40
          Gene Name:
          F-box protein 40
          Gene Aliases:
          9830003A13Rik, mKIAA1195
          Chromosome Location:
          Chr.16: 36963461 - 36990535 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01343539_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxo40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037321.1 NP_001032398.1 2 - 3 2304 84
          GenBank mRNA AK036385.1 - 3 - 4 2062 84
          AK036684.1 - 3 - 4 2059 84
          AK220373.1 - 1 - 2 1938 84
          BC107406.1 - 2 - 3 2304 40058923 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001588.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxo40

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein ubiquitination
          ubiquitin-protein transferase activity
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016307 AK220373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016307 BC107406.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016307 BC107407.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016307 NM_001037321.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016307 AK036385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P279557 AK036684.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P279557 AK036684.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 114295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127626 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128614 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016307 AK220373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016307 BC107406.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016307 BC107407.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016307 NM_001037321.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016307 AK036385.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P279557 AK036684.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P279557 AK036684.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30678363 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08565800 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08565812 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08565832 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128614 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08565834 - -
          Whole Mouse Genome G4122A A_52_P279557 AK036684.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P279557 AK036684.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P297140 AK036385.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P297140 AK036684.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30678346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12985384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12762870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12844430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12626687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12739054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12769217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12626688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12626937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12802240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12541609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12972447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12769216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12860820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12918455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12797811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12843152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12918448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30678327 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10439361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439360 - -
          Affymetrix Mouse Gene 1.1 ST Array 10439357 - -
          Affymetrix Mouse Clariom S Array PSR1600012979.mm.1 NM_001037321.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012979.mm.1 NM_001037321.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012979.mm.1 TC1600001588.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012978.mm.1 NM_001037321.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012978.mm.1 NM_001037321.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012978.mm.1 TC1600001588.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012981.mm.1 NM_001037321.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012981.mm.1 NM_001037321.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012981.mm.1 TC1600001588.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012983.mm.1 NM_001037321.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012983.mm.1 NM_001037321.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012983.mm.1 TC1600001588.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 786335 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2190286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3163206 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3163206 - -
          mouse-6 v1.1 (Illumina) GI_82617543-A - -
          MouseRef-8 v2 Expression BeadChip ILMN_3049851 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3163206 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3125905 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3163206 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3049851 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3125905 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline