Search
Search

Abcc5| Gene Symbol: Abcc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013790.2 | NP_038818.2 | 2 - 3 | 324 | 79 | |
| NM_176839.1 | NP_789809.1 | 2 - 3 | 324 | 79 | ||
| GenBank mRNA | AB019003.1 | - | 2 - 3 | 275 | 79 | |
| AK050565.1 | - | 2 - 3 | 317 | 79 | ||
| BC042581.1 | - | 2 - 3 | 266 | 79 | ||
| BC061132.1 | - | 2 - 3 | 284 | 79 | ||
| BC090629.1 | - | 2 - 3 | 281 | 6839816 | 79 | |
| CX240998.1 | - | 2 - 3 | 300 | 79 | ||
Set Membership: |
|