Search
Search
| Gene Symbol: Ubn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026666.3 | NP_080942.1 | 16 - 17 | 3893 | 87 | |
| GenBank mRNA | AK084046.1 | - | 2 - 3 | 1255 | 87 | |
| AK137727.1 | - | 4 - 5 | 1359 | 87 | ||
| AK147571.1 | - | 16 - 17 | 3894 | 87 | ||
| BC060723.1 | - | 14 - 15 | 2821 | 87 | ||
| BC098372.1 | - | 14 - 15 | 3053 | 30363389 | 87 | |
Set Membership: |
|