Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344021_s1
          See other 2310057N15RIK GE Assays ›
          Gene Symbol:
          2310057N15Rik
          Gene Name:
          RIKEN cDNA 2310057N15 gene
          Gene Aliases:
          Krtap13-1
          Chromosome Location:
          Chr.16: 88773181 - 88774206 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344021_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310057N15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027170.1 NP_081446.1 1 645 178
          GenBank mRNA AK009968.1 - 1 645 178

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2310057N15Rik

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118804 NM_027170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118804 AK009968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118804 BC104340.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118804 BC104341.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159613 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118804 NM_027170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118804 AK009968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118804 BC104340.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118804 BC104341.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947891 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159613 - -
          Whole Mouse Genome G4122A A_51_P193764 NM_027170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193764 AK009968.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102058 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159613 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12676397 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12676407 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12676396 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12676402 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12676404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13009844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12823757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12676391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947900 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440670 - -
          Affymetrix HT MG-430 PM Array Plate 1421575_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10440669 - -
          Mouse Genome 430 2.0 Array 1421575_at - -
          Mouse Genome 430 2.0 Array 1421575_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69696_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69696_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600015525.mm.1 NM_027170.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015525.mm.1 NM_027170.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015525.mm.1 TC1600002013.mm.1 MTA TC
          Mouse Expression Set 430 1421575_at - -
          Mouse Expression Set 430 1421575_PM_at - -
          Mouse Genome 430A 2.0 Array 1421575_at - -
          Mouse Genome 430A 2.0 Array 1421575_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421575_at - -
          Affymetrix HT Mouse Genome 430A Array 1421575_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 633863 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2940025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215300 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215300 - -
          mouse-6 v1.1 (Illumina) scl069696.1_152-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl069696.1_152-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215300 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215300 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline