| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK137010.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK151508.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK035047.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK045752.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK052569.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK137010.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK151508.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK035047.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK045752.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P40954 |
AK052569.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333850 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838500 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
290596 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
732904 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
860696 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
36323 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
243375 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
AK004869.2 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
AK010772.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
AK152412.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
AK163221.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
AK168032.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
AK168163.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
BC004071.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
BC038920.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
BC049925.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
BC055483.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
BC085177.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
BC103800.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
BC117880.2 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
BC117881.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
569199 |
NM_028599.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
114014 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P116233 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P456410 |
AK151508.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P456410 |
AK019912.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK137010.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK151508.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK035047.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK045752.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK052569.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK137010.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK151508.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK035047.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK045752.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P40954 |
AK052569.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P19420 |
AK035047.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P507234 |
AK137010.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P507234 |
AK035047.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P507234 |
AK045752.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P507234 |
AK052569.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P507234 |
NR_028300.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108256 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753225 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753110 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753150 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12616596 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753246 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753163 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753091 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753193 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753201 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753059 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753236 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753107 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12948184 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753090 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753228 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753093 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753149 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753245 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12948183 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753219 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753133 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753205 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753167 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753104 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753239 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12570464 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753096 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753169 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12539761 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753102 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753241 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753055 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753054 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753171 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753178 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753215 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12491577 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753156 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12484416 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753166 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753226 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753212 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12635122 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753097 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753181 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753231 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753174 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12957912 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753158 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12711903 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753184 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12957913 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753153 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753146 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753144 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12957909 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753137 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753217 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753094 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753162 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753145 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12708421 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753214 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12926996 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753089 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753131 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753198 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12708416 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753056 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753235 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12877230 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753195 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12948177 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753190 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753187 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753180 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753206 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12491583 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753243 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12957918 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753200 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753106 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12957926 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753203 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753101 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753147 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753051 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753136 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753152 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12708419 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753108 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753182 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753208 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30753135 |
- |
- |