Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344063_m1
          See other MRPL39 GE Assays ›
          Gene Symbol:
          Mrpl39
          Gene Name:
          mitochondrial ribosomal protein L39
          Gene Aliases:
          C21orf8, MRP-L5, ORF22, Rpml5
          Chromosome Location:
          Chr.16: 84717580 - 84735302 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344063_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl39
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017404.4 NP_059100.3 2 - 3 353 83
          GenBank mRNA AF239728.1 - 2 - 3 270 83
          AK007681.1 - 2 - 3 289 83
          AK031942.1 - 2 - 3 274 83
          BC012274.1 - 2 - 3 232 3981658 83
          BC016561.1 - 2 - 3 289 2645316 83
          BC036276.1 - 2 - 3 289 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001961.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl39

          Panther Classification:

          Molecular Function -

          aminoacyl-tRNA synthetase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial genome maintenance
          translation
          nucleotide binding
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600015181.mm.1 NM_017404.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015181.mm.1 NM_017404.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015181.mm.1 TC1600001961.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015179.mm.1 NM_017404.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015179.mm.1 NM_017404.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015179.mm.1 TC1600001961.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015177.mm.1 NM_017404.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015177.mm.1 NM_017404.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015177.mm.1 TC1600001961.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015176.mm.1 NM_017404.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015176.mm.1 NM_017404.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015176.mm.1 TC1600001961.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015180.mm.1 NM_017404.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015180.mm.1 NM_017404.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015180.mm.1 TC1600001961.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600015183.mm.1 NM_017404.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015183.mm.1 NM_017404.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600015183.mm.1 TC1600001961.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline