Search
Search

Timmdc1| Gene Symbol: Timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024273.2 | NP_077235.2 | 5 - 6 | 727 | 65 | |
| GenBank mRNA | AK081781.1 | - | 5 - 6 | 670 | 65 | |
| BC002243.1 | - | 5 - 6 | 689 | 3492342 | 65 | |
Set Membership: |
|