Search
Search
| Gene Symbol: Pigx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111025.1 | NP_001104495.1 | 3 - 4 | 464 | 98 | |
| NM_024464.3 | NP_077784.2 | 3 - 4 | 470 | 98 | ||
| GenBank mRNA | AK008583.2 | - | 3 - 4 | 472 | 98 | |
| BX525135.1 | - | 3 - 4 | 437 | 98 | ||
| BY708475.1 | - | 3 - 4 | 471 | 98 | ||
| CF581933.1 | - | 3 - 4 | 98 | |||
Set Membership: |
|