Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344766_m1
          See other COX18 GE Assays ›
          Gene Symbol:
          Cox18
          Gene Name:
          cytochrome c oxidase assembly protein 18
          Gene Aliases:
          BC038311
          Chromosome Location:
          Chr.5: 90214724 - 90223996 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344766_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033310.3 NP_001028482.2 2 - 3 479 64
          NM_001163456.1 NP_001156928.1 2 - 3 479 64
          NR_028088.1 2 - 3 483 64
          GenBank mRNA AK156053.1 - 2 - 3 448 64
          BC038311.1 - 2 - 3 249 64
          BC138972.1 - 2 - 3 457 8861994 64
          BC145479.1 - 2 - 3 457 9054042 64
          BC145486.1 - 2 - 3 461 64
          BC145728.1 - 2 - 3 457 40131031 64
          CX225681.1 - 2 - 3 406 64
          DN174415.1 - 2 - 3 454 64
          DV661727.1 - 1 - 2 90 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002722.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox18

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          respiratory chain complex IV assembly
          protein transport
          protein insertion into mitochondrial membrane
          protein insertion into membrane
          protein transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881743 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881744 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881742 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881745 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990134 BE690958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990134 AK034738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990134 AK156053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990134 BC021612.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990134 BC038311.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4536 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173244 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990134 BE690958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990134 AK034738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990134 AK156053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990134 BC021612.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990134 BC038311.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23881753 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173244 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05650757 - -
          Whole Mouse Genome G4122A A_52_P665767 BE690958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665767 AK034738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665767 NM_001033310.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665767 NM_001163456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665767 NR_028088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665768 BE690958.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665768 AK034738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665768 NM_001033310.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665768 NM_001163456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P665768 NR_028088.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138387 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173244 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881743 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881742 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881744 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23881745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23881756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04209146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04152953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04879502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04430805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04430796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04710313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04430795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04430812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04755597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04419730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23881734 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10531212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531211 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531209 - -
          Affymetrix HT MG-430 PM Array Plate 1415710_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10531208 - -
          Mouse Genome 430 2.0 Array 1415710_at - -
          Mouse Genome 430 2.0 Array 1415710_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231430_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 231430_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500024008.mm.1 NM_001033310.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500024008.mm.1 NM_001033310.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500024008.mm.1 NM_001163456.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500024008.mm.1 NM_001163456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500024008.mm.1 NR_028088.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500024008.mm.1 NR_028088.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500024008.mm.1 TC0500002722.mm.1 MTA TC
          Mouse Expression Set 430 1415710_PM_at - -
          Mouse Expression Set 430 1415710_at - -
          Mouse Expression Set 430 1415710_PM_at - -
          Mouse Expression Set 430 1415710_at - -
          Murine Genome U74v2 Set 162029_r_at - -
          Murine Genome U74v2 Set 92388_at - -
          Murine Genome U74v2 Set 92388_PM_at - -
          Murine Genome U74v2 Set 162029_PM_r_at - -
          Murine Genome U74v2 Set 170220_PM_r_at - -
          Murine Genome U74v2 Set 170220_r_at - -
          Mouse Genome 430A 2.0 Array 1415710_at - -
          Mouse Genome 430A 2.0 Array 1415710_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415710_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415710_at - -
          Affymetrix HT Mouse Genome 430B Array 1415710_at - -
          Affymetrix HT Mouse Genome 430B Array 1415710_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 912436 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2633039 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3002011 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3002011 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline