Search
Search

Frmpd1| Gene Symbol: Frmpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081172.2 | NP_001074641.1 | 6 - 7 | 569 | 72 | |
| GenBank mRNA | AK122409.1 | - | 3 - 4 | 195 | 72 | |
| BC158060.1 | - | 6 - 7 | 533 | 9007360 | 72 | |
Set Membership: |
|