Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm01345342_m1
See other E230008N13RIK GE Assays ›
Gene Symbol:
E230008N13Rik
Gene Name:
RIKEN cDNA E230008N13 gene
Gene Aliases:
Gm1025
Chromosome Location:
Chr.4: 45890303 - 45950774 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm01345342_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: E230008N13Rik
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_198660.2
NP_941062.2
31 - 32
4102
65
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC0400000497.mm.1
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Recommended Assay
Mus musculus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene E230008N13Rik
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
molecular_function
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR0400003572.mm.1
NM_198660.2.1
Refseq
Affymetrix Mouse Clariom S Array
PSR0400003572.mm.1
NM_198660.2
Refseq
Affymetrix Mouse Clariom S Array
PSR0400003572.mm.1
TC0400000497.mm.1
MTA TC
Affymetrix Mouse Clariom S Array
PSR0400003571.mm.1
NM_198660.2.1
Refseq
Affymetrix Mouse Clariom S Array
PSR0400003571.mm.1
NM_198660.2
Refseq
Affymetrix Mouse Clariom S Array
PSR0400003571.mm.1
TC0400000497.mm.1
MTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
git-commit: c9e08c96761173abe34e68f880379696776a4827
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.1-2026.02.97.1.0