Search
Search

Tmod1| Gene Symbol: Tmod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021883.2 | NP_068683.1 | 5 - 6 | 621 | 61 | |
| GenBank mRNA | AK132878.1 | - | 4 - 5 | 454 | 61 | |
| AK177919.1 | - | 2 - 3 | 157 | 61 | ||
| AK216897.1 | - | 2 - 3 | 157 | 61 | ||
| BC106849.1 | - | 4 - 5 | 509 | 40046092 | 61 | |
| BC106850.1 | - | 4 - 5 | 509 | 40046093 | 61 | |
| S76831.1 | - | 4 - 5 | 548 | 61 | ||
Set Membership: |
|