Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01346299_s1
          See other OLFR195 GE Assays ›
          Gene Symbol:
          Olfr195
          Gene Name:
          olfactory receptor 195
          Gene Aliases:
          MOR184-5
          Chromosome Location:
          Chr.16: 59131709 - 59149817 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01346299_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr195
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146998.1 NP_667209.1 1 818 99
          GenBank mRNA BC119110.1 - 1 841 8733859 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr195

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065459 NM_146998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2065459 BC119110.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164918 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065459 NM_146998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2065459 BC119110.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251684 NM_146998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251684 BC119110.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794512 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164918 - -
          Whole Mouse Genome G4122A A_51_P251684 NM_146998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251684 BC119110.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111879 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12487893 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12674618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12857816 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12857818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12487891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12932410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12674615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12860539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12627485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13049395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12913420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12674636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794512 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10436413 - -
          Affymetrix Mouse Gene 1.1 ST Array 10436412 - -
          Affymetrix Mouse Clariom S Array PSR1600006669.mm.1 TC1600000768.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 883531 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7100494 - -
          Illumina mouse-6 v1.0 expression beadchip 7100494 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100494 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247436 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247436 - -
          mouse-6 v1.1 (Illumina) scl1217.1.1_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100494 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl1217.1.1_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100494 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247436 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247436 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline