Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01346473_sH
          See other OLFR207 GE Assays ›
          Gene Symbol:
          Olfr207
          Gene Name:
          olfactory receptor 207
          Gene Aliases:
          MOR182-9
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01346473_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr207
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011792.1 NP_001011792.1 1 329 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001802.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr207

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2122117 NM_001011792.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141321 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2122117 NM_001011792.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30795242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30795239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30795246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30795235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30795241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30795245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30795244 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141321 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141321 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12988071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30795230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12902821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12909573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12988071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30795230 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440181 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440165 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440184 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440180 - -
          Affymetrix Mouse Clariom S Array PSR1600014483.mm.1 NM_207551.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014483.mm.1 NM_207551.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014483.mm.1 TC1600001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600014484.mm.1 NM_207551.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014484.mm.1 NM_207551.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014484.mm.1 TC1600001802.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 599522 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_3007198 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3007198 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline