Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01348077_m1
          See other PGLYRP2 GE Assays ›
          Gene Symbol:
          Pglyrp2
          Gene Name:
          peptidoglycan recognition protein 2
          Gene Aliases:
          C730002N09Rik, PGRP-L, Pglyrpl, tagL
          Chromosome Location:
          Chr.17: 32412459 - 32424183 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01348077_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pglyrp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271476.1 NP_001258405.1 2 - 3 284 88
          NM_001271477.1 NP_001258406.1 1 - 2 176 88
          NM_001271478.1 NP_001258407.1 2 - 3 284 88
          NM_001271479.1 NP_001258408.1 1 - 2 176 88
          NM_021319.5 NP_067294.2 1 - 2 176 88
          NR_073184.1 1 - 2 176 88
          GenBank mRNA AB084140.1 - 2 - 3 284 88
          AB084141.1 - 2 - 3 284 88
          AF149837.1 - 1 - 2 176 88
          AF149838.1 - 1 - 2 176 88
          AF315578.1 - 1 - 2 176 88
          AF315584.1 - 1 - 2 176 88
          AF317548.1 - 1 - 2 176 88
          AF392055.1 - 2 - 3 210 88
          AK039953.1 - 2 - 3 284 88
          AK050019.1 - 1 - 2 177 88
          AY282722.1 - 1 - 2 114 88
          BC019396.1 - 1 - 2 149 5124925 88
          EF204951.1 - 1 - 2 64 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001846.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pglyrp2

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern recognition receptor signaling pathway
          immune system process
          peptidoglycan catabolic process
          detection of bacterium
          negative regulation of interferon-gamma production
          negative regulation of natural killer cell differentiation involved in immune response
          growth of symbiont in host
          regulation of inflammatory response
          defense response to Gram-positive bacterium
          zinc ion binding
          N-acetylmuramoyl-L-alanine amidase activity
          peptidoglycan receptor activity
          hydrolase activity
          peptidoglycan binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700017096.mm.1 NM_001271476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017096.mm.1 NM_001271477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017096.mm.1 NM_001271478.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017096.mm.1 NM_001271479.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017096.mm.1 NM_021319.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017096.mm.1 NR_073184.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017096.mm.1 TC1700001846.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017098.mm.1 NM_001271477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017098.mm.1 NM_001271479.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017098.mm.1 NM_021319.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017098.mm.1 NR_073184.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017098.mm.1 TC1700001846.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017099.mm.1 NM_001271477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017099.mm.1 NM_001271479.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017099.mm.1 NM_021319.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017099.mm.1 NR_073184.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017099.mm.1 TC1700001846.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 666797 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline