Search
Search

Vmn1r21| Gene Symbol: Vmn1r21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134183.1 | NP_598944.1 | 1 | 48 | 102 | |
| GenBank mRNA | BC120799.1 | - | 1 | 89 | 40129722 | 102 |
| BC120801.1 | - | 1 | 89 | 40129724 | 102 | |
Set Membership: |
|