Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01348792_m1
          See other 2700029M09RIK GE Assays ›
          Gene Symbol:
          2700029M09Rik
          Gene Name:
          RIKEN cDNA 2700029M09 gene
          Gene Aliases:
          C230006B22Rik
          Chromosome Location:
          Chr.8: 60890393 - 60907582 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01348792_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2700029M09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028299.1 NP_082575.1 7 - 8 947 73
          GenBank mRNA AK012304.1 - 7 - 8 920 73
          AK150453.1 - 7 - 8 933 73
          AK151754.1 - 6 - 7 764 73
          AK157260.1 - 7 - 8 933 73
          BC042740.1 - 7 - 8 947 5024951 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000648.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906691 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906690 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906687 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906689 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906688 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 BC042740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 AK012304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 AK150453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 AK151754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 AK157260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 BC042740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 AK012304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 AK150453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 AK151754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450518 AK157260.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76082 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157078 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112244 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 BC042740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 AK012304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 AK150453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 AK151754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 AK157260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 BC042740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 AK012304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 AK150453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 AK151754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450518 AK157260.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112244 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06529105 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157078 - -
          Whole Mouse Genome G4122A A_51_P450518 BC042740.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450518 AK012304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450518 AK150453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450518 AK151754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450518 AK157260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450518 BC042740.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450518 AK012304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450518 AK150453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450518 AK151754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450518 AK157260.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112244 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157078 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906687 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906688 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906689 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906690 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25906691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07352735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06729511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06952751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06952748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07029226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07001503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06836890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25906707 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10571912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571918 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571916 - -
          Affymetrix Mouse Gene 1.0 ST Array 10571919 - -
          Affymetrix HT MG-430 PM Array Plate 1452877_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10571911 - -
          Mouse Genome 430 2.0 Array 1452877_PM_at - -
          Mouse Genome 430 2.0 Array 1452877_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72612_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72612_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800004328.mm.1 NM_028299.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004328.mm.1 NM_028299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004328.mm.1 TC0800000648.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800004330.mm.1 NM_028299.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004330.mm.1 NM_028299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800004330.mm.1 TC0800000648.mm.1 MTA TC
          Mouse Expression Set 430 1452877_at - -
          Mouse Expression Set 430 1452877_PM_at - -
          Murine Genome U74v2 Set 94360_PM_at - -
          Murine Genome U74v2 Set 94360_at - -
          Mouse Genome 430A 2.0 Array 1452877_at - -
          Mouse Genome 430A 2.0 Array 1452877_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452877_at - -
          Affymetrix HT Mouse Genome 430A Array 1452877_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253477 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253477 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline