| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400032506.mm.1 |
TC0400003911.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032506.mm.1 |
NM_001081355.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032506.mm.1 |
NM_001081355.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032506.mm.1 |
NM_001256380.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032506.mm.1 |
NM_001256380.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032508.mm.1 |
TC0400003911.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032508.mm.1 |
NM_001081355.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032508.mm.1 |
NM_001081355.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032508.mm.1 |
NM_001256380.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032508.mm.1 |
NM_001256380.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032509.mm.1 |
TC0400003911.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032509.mm.1 |
NM_001081355.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032509.mm.1 |
NM_001081355.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032509.mm.1 |
NM_001256380.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032509.mm.1 |
NM_001256380.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032507.mm.1 |
TC0400003911.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032507.mm.1 |
NM_001081355.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032507.mm.1 |
NM_001081355.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032507.mm.1 |
NM_001256380.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032507.mm.1 |
NM_001256380.1 |
Refseq |