| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800005460.mm.1 |
NM_001166399.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005460.mm.1 |
NM_001166399.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005460.mm.1 |
NM_009001.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005460.mm.1 |
NM_009001.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005460.mm.1 |
TC0800000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800005463.mm.1 |
NM_001166399.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005463.mm.1 |
NM_001166399.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005463.mm.1 |
NM_009001.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005463.mm.1 |
NM_009001.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005463.mm.1 |
TC0800000765.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800005458.mm.1 |
NM_001166399.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005458.mm.1 |
NM_001166399.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005458.mm.1 |
NM_009001.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005458.mm.1 |
NM_009001.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800005458.mm.1 |
TC0800000765.mm.1 |
MTA TC |