Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01349242_g1
          See other PPP3R2 GE Assays ›
          Gene Symbol:
          Ppp3r2
          Gene Name:
          protein phosphatase 3, regulatory subunit B, alpha isoform (calcineurin B, type II)
          Gene Aliases:
          CaNB2, CnB2
          Chromosome Location:
          Chr.4: 49678747 - 49681983 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01349242_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp3r2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004025.4 NP_001004025.1 1 - 2 549 100
          GenBank mRNA AK029998.1 - 1 - 2 551 100
          AK132760.1 - 1 - 2 574 100
          AK161263.1 - 1 - 2 551 100
          BC100409.1 - 1 - 2 583 30919135 100
          S43865.1 - 1 - 2 580 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002643.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp3r2

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen metabolic process
          defense response
          sperm displacement
          dephosphorylation
          calcium-dependent protein serine/threonine phosphatase activity
          calcium ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031352 AK029998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031352 NM_001004025.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141755 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2031352 AK029998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2031352 NM_001004025.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136814 S43865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136814 AK029998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136814 AK132760.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136814 AK161263.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136814 BC100409.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136810 S43865.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136810 AK029998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136810 AK132760.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136810 AK161263.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P136810 BC100409.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22906445 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141755 - -
          Whole Mouse Genome G4122A A_51_P136814 S43865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136814 AK029998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136814 AK132760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136814 AK161263.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136814 BC100409.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136810 S43865.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136810 AK029998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136810 AK132760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136810 AK161263.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136810 BC100409.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141755 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03242947 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03242956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03242923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03242929 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03242949 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03242954 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03242934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22906437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03273737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03242938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03242921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04053048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03524609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03242937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03358787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03391654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03242970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04053050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04053047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03524605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22906523 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512931 - -
          Affymetrix HT MG-430 PM Array Plate 1437535_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512930 - -
          Mouse Genome 430 2.0 Array 1437535_PM_at - -
          Mouse Genome 430 2.0 Array 1437535_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19059_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19059_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400021538.mm.1 TC0400002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021538.mm.1 NM_001004025.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021538.mm.1 NM_001004025.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021539.mm.1 TC0400002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021537.mm.1 TC0400002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021537.mm.1 NM_001004025.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021537.mm.1 NM_001004025.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021536.mm.1 TC0400002643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021536.mm.1 NM_001004025.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021536.mm.1 NM_001004025.4 Refseq
          Mouse Expression Set 430 1437535_at - -
          Mouse Expression Set 430 1437535_PM_at - -
          Murine Genome U74v2 Set 165481_PM_at - -
          Murine Genome U74v2 Set 165481_at - -
          Affymetrix HT Mouse Genome 430B Array 1437535_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437535_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 644239 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2592092 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2592092 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline