Search
Search

Rusc2| Gene Symbol: Rusc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037709.2 | NP_001032798.1 | 5 - 6 | 3280 | 61 | |
| NM_199057.3 | NP_951012.2 | 5 - 6 | 3241 | 61 | ||
| GenBank mRNA | AK053542.1 | - | 2 - 3 | 1542 | 61 | |
| AK122263.1 | - | 5 - 6 | 3160 | 61 | ||
| AK132170.1 | - | 5 - 6 | 3170 | 61 | ||
| AK166260.1 | - | 5 - 6 | 3181 | 61 | ||
| BC024790.1 | - | 3 - 4 | 471 | 61 | ||
| BC141186.1 | - | 4 - 5 | 3011 | 9055798 | 61 | |
Set Membership: |
|