Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600004964.mm.1 |
NM_030035.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004964.mm.1 |
NM_030035.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004964.mm.1 |
TC1600000528.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004960.mm.1 |
NM_030035.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004960.mm.1 |
NM_030035.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004960.mm.1 |
TC1600000528.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004962.mm.1 |
NM_030035.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004962.mm.1 |
NM_030035.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004962.mm.1 |
TC1600000528.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004963.mm.1 |
NM_030035.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004963.mm.1 |
NM_030035.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004963.mm.1 |
TC1600000528.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004959.mm.1 |
NM_030035.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004959.mm.1 |
NM_030035.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004959.mm.1 |
TC1600000528.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004957.mm.1 |
NM_030035.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004957.mm.1 |
NM_030035.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004957.mm.1 |
TC1600000528.mm.1 |
MTA TC |