Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01349855_s1
          See other OLFR198 GE Assays ›
          Gene Symbol:
          Olfr198
          Gene Name:
          olfactory receptor 198
          Gene Aliases:
          MOR182-8
          Chromosome Location:
          Chr.16: 59201504 - 59202424 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01349855_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr198
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011808.1 NP_001011808.1 1 878 114
          GenBank mRNA BC147215.1 - 1 973 9056251 114

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr198

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959328 BC147215.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959328 NM_001011808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959328 BC147215.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959328 NM_001011808.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30794683 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08614001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30794678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13036393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12638125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30794681 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440171 - -
          Affymetrix Mouse Gene 1.1 ST Array 10440170 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 900302 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160318 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160318 - -
          mouse-6 v1.1 (Illumina) GI_58801381-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160318 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline