Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01350720_m1
          See other KLHL40 GE Assays ›
          Gene Symbol:
          Klhl40
          Gene Name:
          kelch-like 40
          Gene Aliases:
          2310024D23Rik, Kbtbd5
          Chromosome Location:
          Chr.9: 121777101 - 121791002 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01350720_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klhl40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028202.3 NP_082478.1 4 - 5 1779 75
          GenBank mRNA AK009491.1 - 4 - 5 1695 75
          AK078669.1 - 4 - 5 1693 75
          AK160799.1 - 4 - 5 1639 75

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          negative regulation of protein ubiquitination
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          skeletal muscle fiber development
          skeletal muscle fiber differentiation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885524 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885523 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885527 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885525 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885528 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885522 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885526 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235139 AK160799.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235139 NM_028202.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235139 AK009491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235139 AK078669.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235139 AK160799.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235139 NM_028202.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235139 AK009491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P235139 AK078669.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292105 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235139 AK160799.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235139 NM_028202.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235139 AK009491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235139 AK078669.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235139 AK160799.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235139 NM_028202.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235139 AK009491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P235139 AK078669.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235139 AK160799.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235139 NM_028202.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235139 AK009491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235139 AK078669.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235139 AK160799.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235139 NM_028202.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235139 AK009491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P235139 AK078669.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108216 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133781 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885522 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885524 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885528 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885526 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885523 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885527 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08008481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07603857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07813438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07586505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07923135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08041420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08154120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07813433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07923130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07603856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07969077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07586489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07586494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08154121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07586493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07969072 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10590423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590425 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590421 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590420 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590426 - -
          Affymetrix HT MG-430 PM Array Plate 1431043_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10590417 - -
          Mouse Genome 430 2.0 Array 1431043_at - -
          Mouse Genome 430 2.0 Array 1431043_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72330_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72330_PM_at - -
          Mouse Expression Set 430 1431043_at - -
          Mouse Expression Set 430 1431043_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431043_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431043_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 340532 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2636054 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2636054 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline