Search
Search

Ppp1r3fos| Gene Symbol: Ppp1r3fos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019695.1 | - | 1 - 2 | 112 | 103 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein phosphatase 1, regulatory (inhibitor) subunit 3F |
Ppp1r3f |
DXImx48e, R3F, RF3, Sfc15 |
-211.0 | Anti-sense | Chr. X - 7558562 - 7574281 on NCBI Build GRCm38 |
| Downstream | forkhead box P3 | Foxp3 |
JM2, scurfin, sf |
2.0kb | Sense | Chr. X - 7579676 - 7595243 on NCBI Build GRCm38 |
Set Membership: |
|