Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01350975_g1
          See other 4933432G23RIK GE Assays ›
          Gene Symbol:
          4933432G23Rik
          Gene Name:
          RIKEN cDNA 4933432G23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 118429839 - 118437048 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01350975_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933432G23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040716.1 4 - 5 571 69
          GenBank mRNA AK017021.1 - 4 - 5 575 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001560.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84804 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141741 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141741 - -
          Whole Mouse Genome G4122A A_52_P753860 AK017021.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133899 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07776615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07776616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07678598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08085298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07546476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07898869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07898864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07678597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07812741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26866323 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900012914.mm.1 NR_040716.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900012914.mm.1 NR_040716.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900012914.mm.1 TC0900001560.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 300067 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107100324 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline