Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01351463_s1
          See other OLFR155 GE Assays ›
          Gene Symbol:
          Olfr155
          Gene Name:
          olfactory receptor 155
          Gene Aliases:
          GA_x5J8B7W5BNN-979337-980296, MOR262-14, OR37A, Olfr37a, mOR37a
          Chromosome Location:
          Chr.4: 43854292 - 43855463 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01351463_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr155
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019473.1 NP_062346.1 1 1033 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr155

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P305600 NM_019473.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P305600 NM_019473.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120730 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P305600 NM_019473.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P305600 NM_019473.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P305600 NM_019473.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P305600 NM_019473.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873946 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120730 - -
          Whole Mouse Genome G4122A A_51_P305600 NM_019473.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P305600 NM_019473.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124611 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22874019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22874008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04057385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03562115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03599017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03921067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03921078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03921069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04057383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04059478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03599021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03897203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03764850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03814764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03897205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03341623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03599019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22874004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22874007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03921074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03946667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03562118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873937 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10504423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512631 - -
          Affymetrix HT MG-430 PM Array Plate 1422343_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10504422 - -
          Affymetrix Mouse Gene 1.1 ST Array 10512630 - -
          Mouse Genome 430 2.0 Array 1422343_PM_at - -
          Mouse Genome 430 2.0 Array 1422343_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 29845_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 29845_at - -
          Mouse Expression Set 430 1422343_at - -
          Mouse Expression Set 430 1422343_PM_at - -
          Murine Genome U74v2 Set 96999_at - -
          Murine Genome U74v2 Set 96999_PM_at - -
          Mouse Genome 430A 2.0 Array 1422343_at - -
          Mouse Genome 430A 2.0 Array 1422343_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422343_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422343_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 320219 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4150156 - -
          Illumina mouse-6 v1.0 expression beadchip 4150156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150156 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720678 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720678 - -
          mouse-6 v1.1 (Illumina) scl25504.1.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150156 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25504.1.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2720678 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2720678 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0