Search
Search
| Gene Symbol: Dek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025900.2 | NP_080176.2 | 4 - 5 | 525 | 142 | |
| GenBank mRNA | AK012615.1 | - | 4 - 5 | 466 | 142 | |
| AK012877.1 | - | 4 - 5 | 460 | 142 | ||
| AK033451.1 | - | 4 - 5 | 415 | 142 | ||
| AK154013.1 | - | 4 - 5 | 507 | 142 | ||
| BC048844.1 | - | 4 - 5 | 475 | 142 | ||
| BC055451.1 | - | 4 - 5 | 471 | 4164972 | 142 | |
Set Membership: |
|