Search
Search

4933417A18Rik| Gene Symbol: 4933417A18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025750.3 | NP_080026.2 | 1 - 2 | 305 | 124 | |
| GenBank mRNA | AK016778.1 | - | 1 - 2 | 270 | 124 | |
| AK016838.1 | - | 1 - 2 | 306 | 124 | ||
| BC100450.1 | - | 1 - 2 | 295 | 30919421 | 124 | |
| BC145747.1 | - | 1 - 2 | 260 | 40131050 | 124 | |
Set Membership: |
|