| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
435226 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
714992 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
600504 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905374 |
AK005163.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905374 |
AK007686.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905374 |
AK168868.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905374 |
BC012515.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905374 |
BC030487.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
905374 |
NM_028058.3 |
refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01761802 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07180530 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947617 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947620 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947608 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07180523 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947607 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947641 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06891359 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07280965 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07040493 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06994225 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947609 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947623 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947648 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947626 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947615 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947644 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07107953 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07084069 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947627 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947632 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947621 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947603 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06698919 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07040498 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947631 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25947628 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06994226 |
- |
- |