Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01352763_g1
          See other TCHHL1 GE Assays ›
          Gene Symbol:
          Tchhl1
          Gene Name:
          trichohyalin-like 1
          Gene Aliases:
          5430400H23Rik, AA589420, S100a17, Thhl1
          Chromosome Location:
          Chr.3: 93468734 - 93471980 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01352763_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tchhl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027762.3 NP_082038.2 2 - 3 216 87
          GenBank mRNA AA856087.1 - 1 - 2 86 87
          AK017244.1 - 2 - 3 217 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000850.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tchhl1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038242 AK017244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038242 NM_027762.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 570490 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127589 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038242 AK017244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038242 NM_027762.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22336514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127589 - -
          Whole Mouse Genome G4122A A_51_P473912 AK017244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P473912 NM_027762.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132635 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03007552 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03007564 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03007550 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03007559 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03007547 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03007571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22336541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03007569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03027031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02304591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03019249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02801526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02801524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02718313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03170605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02718311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02718316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03007560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02718295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02951620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02672470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22336507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02718304 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493989 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493988 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493987 - -
          Affymetrix HT MG-430 PM Array Plate 1429944_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493986 - -
          Mouse Genome 430 2.0 Array 1429944_PM_at - -
          Mouse Genome 430 2.0 Array 1429944_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71325_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71325_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300006528.mm.1 NM_027762.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006528.mm.1 NM_027762.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006528.mm.1 TC0300000850.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006527.mm.1 NM_027762.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006527.mm.1 NM_027762.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006527.mm.1 TC0300000850.mm.1 MTA TC
          Mouse Expression Set 430 1429944_at - -
          Mouse Expression Set 430 1429944_PM_at - -
          Murine Genome U74v2 Set 169938_PM_at - -
          Murine Genome U74v2 Set 169938_at - -
          Affymetrix HT Mouse Genome 430B Array 1429944_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429944_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 679975 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5720072 - -
          Illumina mouse-6 v1.0 expression beadchip 5720072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2718552 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2718552 - -
          mouse-6 v1.1 (Illumina) scl22911.3_700-S - -
          Illumina Sentrix mouse-6.v1 bead array 5720072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22911.3_700-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2718552 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2718552 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline