Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01352860_m1
          See other LYSMD3 GE Assays ›
          Gene Symbol:
          Lysmd3
          Gene Name:
          LysM, putative peptidoglycan-binding, domain containing 3
          Gene Aliases:
          1110030H10Rik, BC003322
          Chromosome Location:
          Chr.13: 81657657 - 81672871 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01352860_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lysmd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030257.1 NP_084533.1 2 - 3 397 152
          GenBank mRNA BC003322.1 - 2 - 3 397 3156298 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000965.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lysmd3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177063 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177065 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177062 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177061 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177064 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91618 AK029514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91618 BC003322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91618 NM_030257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91618 AK029514.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91618 BC003322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P91618 NM_030257.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 259756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770346 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69610 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 557917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149333 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170636 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100319 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91618 AK029514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91618 BC003322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91618 NM_030257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91618 AK029514.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91618 BC003322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P91618 NM_030257.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P214573 AK003980.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29177057 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170636 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100319 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07933799 - -
          Whole Mouse Genome G4122A A_52_P91618 AK029514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91618 BC003322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91618 NM_030257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91618 AK029514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91618 BC003322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P91618 NM_030257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214573 AK003980.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133690 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100319 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170636 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10594925 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11120006 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11120008 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10627473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11096707 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177064 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177062 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177061 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177065 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29177063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29177105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10594925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10633129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11052315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10906564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11120008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10858568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10906568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11052311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10932752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29177043 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10406422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10406420 - -
          Affymetrix Mouse Gene 1.0 ST Array 10406421 - -
          Affymetrix HT MG-430 PM Array Plate 1460335_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434174_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10406419 - -
          Mouse Genome 430 2.0 Array 1460335_PM_at - -
          Mouse Genome 430 2.0 Array 1460335_at - -
          Mouse Genome 430 2.0 Array 1434174_PM_at - -
          Mouse Genome 430 2.0 Array 1434174_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80289_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80289_at - -
          Affymetrix Mouse Clariom S Array PSR1300006220.mm.1 NM_030257.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006220.mm.1 NM_030257.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006220.mm.1 TC1300000965.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300006221.mm.1 NM_030257.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006221.mm.1 NM_030257.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006221.mm.1 TC1300000965.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300006222.mm.1 NM_030257.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006222.mm.1 NM_030257.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006222.mm.1 TC1300000965.mm.1 MTA TC
          Mouse Expression Set 430 1460335_PM_at - -
          Mouse Expression Set 430 1460335_at - -
          Mouse Expression Set 430 1434174_at - -
          Mouse Expression Set 430 1434174_PM_at - -
          Murine Genome U74v2 Set 168018_PM_at - -
          Murine Genome U74v2 Set 168018_at - -
          Mouse Genome 430A 2.0 Array 1460335_at - -
          Mouse Genome 430A 2.0 Array 1460335_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460335_at - -
          Affymetrix HT Mouse Genome 430A Array 1460335_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434174_at - -
          Affymetrix HT Mouse Genome 430B Array 1434174_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 554972 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1450114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2642489 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2642489 - -
          mouse-6 v1.1 (Illumina) scl44581.3_458-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44581.3_458-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2642489 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2642489 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline