Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01354211_s1
          See other OLFR324 GE Assays ›
          Gene Symbol:
          Olfr324
          Gene Name:
          olfactory receptor 324
          Gene Aliases:
          MOR102-2
          Chromosome Location:
          Chr.11: 58597345 - 58598466 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01354211_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr324
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011743.2 NP_001011743.2 1 1010 77
          GenBank mRNA BC147575.1 - 1 1010 9056565 77
          BC147579.1 - 1 1010 9056569 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr324

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980671 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27831748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09192761 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09192798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09192771 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09192788 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09192782 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09192760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27831754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09202951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09026157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09257670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09257676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09026161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09520476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09192798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09192794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09192801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09202960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27831792 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376395 - -
          Affymetrix Mouse Gene 1.1 ST Array 10376394 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 459846 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161220 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161220 - -
          mouse-6 v1.1 (Illumina) GI_58801279-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161220 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161220 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7mgp5:80/100.66.78.208:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0