Search
Search

Fam172a| Gene Symbol: Fam172a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163419.1 | NP_001156891.1 | 6 - 7 | 850 | 80 | |
| NM_001163420.1 | NP_001156892.1 | 5 - 6 | 658 | 80 | ||
| NM_138312.1 | NP_612185.1 | 7 - 8 | 974 | 80 | ||
| NR_028109.1 | 7 - 8 | 899 | 80 | |||
| GenBank mRNA | AF064782.1 | - | 6 - 7 | 803 | 80 | |
| AK052419.1 | - | 6 - 7 | 822 | 80 | ||
| AK165273.1 | - | 7 - 8 | 873 | 80 | ||
| AK166285.1 | - | 5 - 6 | 630 | 80 | ||
| BC057380.1 | - | 6 - 7 | 818 | 80 | ||
Set Membership: |
|