Search
Search

Arl13a| Gene Symbol: Arl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028947.1 | NP_083223.1 | 3 - 4 | 304 | 71 | |
| GenBank mRNA | AK016872.1 | - | 3 - 4 | 305 | 71 | |
| BC100522.1 | - | 3 - 4 | 202 | 30920038 | 71 | |
Set Membership: |
|