Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01545236_m1
          See other PID1 GE Assays ›
          Gene Symbol:
          Pid1
          Gene Name:
          phosphotyrosine interaction domain containing 1
          Gene Aliases:
          5033414K04Rik, NYGGF4
          Chromosome Location:
          Chr.1: 84036293 - 84284675 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01545236_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pid1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001003948.2 NP_001003948.2 1 - 2 309 96
          GenBank mRNA AK035542.1 - 1 - 2 184 96
          AK083256.1 - 1 - 2 229 96
          AK150971.1 - 1 - 2 247 96
          AK151267.1 - 1 - 2 279 96
          AK164328.1 - 1 - 2 276 96
          BC080290.1 - 1 - 2 255 6816865 96

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC080290.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pid1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          positive regulation of gene expression
          cellular response to leptin stimulus
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          regulation of mitochondrial membrane potential
          positive regulation of fat cell proliferation
          mitochondrion morphogenesis
          cellular response to cytokine stimulus
          cellular response to interleukin-6
          cellular response to tumor necrosis factor
          cellular response to fatty acid
          negative regulation of establishment of protein localization to plasma membrane
          negative regulation of mitochondrial DNA replication
          regulation of G1/S transition of mitotic cell cycle
          regulation of reactive oxygen species metabolic process
          positive regulation of reactive oxygen species metabolic process
          negative regulation of ATP biosynthetic process
          positive regulation of ATP biosynthetic process
          negative regulation of glucose import in response to insulin stimulus
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100021834.mm.1 NM_001003948.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021834.mm.1 NM_001003948.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021834.mm.1 TC0100002690.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100021835.mm.1 NM_001003948.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021835.mm.1 NM_001003948.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021835.mm.1 TC0100002690.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100021832.mm.1 NM_001003948.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021832.mm.1 NM_001003948.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021832.mm.1 TC0100002690.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100021837.mm.1 NM_001003948.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021837.mm.1 NM_001003948.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100021837.mm.1 TC0100002690.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 471272 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline