Search
Search

Fam179a| Gene Symbol: Fam179a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177087.4 | NP_796061.2 | 11 - 12 | 1861 | 55 | |
| GenBank mRNA | AK076284.1 | - | 10 - 11 | 1515 | 55 | |
| AK158647.1 | - | 11 - 12 | 1646 | 55 | ||
Set Membership: |
|