Search
Search
| Gene Symbol: Rhoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016802.4 | NP_058082.2 | 1 - 2 | 345 | 92 | |
| GenBank mRNA | AF014371.1 | - | 53 | 91 | ||
| AK077606.1 | - | 1 - 2 | 153 | 92 | ||
| AK083624.1 | - | 1 - 2 | 152 | 92 | ||
| AK150294.1 | - | 1 - 2 | 190 | 92 | ||
| AK150475.1 | - | 1 - 2 | 196 | 92 | ||
| AK152629.1 | - | 1 - 2 | 132 | 92 | ||
| AK165512.1 | - | 1 - 2 | 128 | 92 | ||
| AK172051.1 | - | 1 - 2 | 153 | 92 | ||
| BC068115.1 | - | 1 - 2 | 345 | 6809149 | 92 | |
| BC096423.1 | - | 1 - 2 | 131 | 3590185 | 92 | |
Set Membership: |
|