Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01603599_g1
          See other OLFR611 GE Assays ›
          Gene Symbol:
          Olfr611
          Gene Name:
          olfactory receptor 611
          Gene Aliases:
          MOR15-1, MOR15-4
          Chromosome Location:
          Chr.7: 103517411 - 103518382 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01603599_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr611
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146727.2 NP_666938.2 1 546 127

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr611

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964528 NM_001200027.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964528 NM_146727.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163398 NM_146727.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163398 NM_146727.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P129972 NM_146727.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25383682 - -
          Whole Mouse Genome G4122A A_51_P129972 NM_146727.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06219220 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06219222 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06219263 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06219247 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06219257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25383598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06219245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25383683 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566222 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566223 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566221 - -
          Affymetrix Mouse Clariom S Array PSR0700032719.mm.1 NM_146727.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032719.mm.1 NM_146727.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032719.mm.1 TC0700003894.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2350471 - -
          Illumina mouse-6 v1.0 expression beadchip 2350471 - -
          Illumina mouse-6 v1.0 expression beadchip 5670722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2735071 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253092 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253092 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2735071 - -
          mouse-6 v1.1 (Illumina) scl0258722.1_9-S - -
          mouse-6 v1.1 (Illumina) scl00258722.1_67-S - -
          Illumina Sentrix mouse-6.v1 bead array 5670722 - -
          Illumina Sentrix mouse-6.v1 bead array 2350471 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258722.1_9-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258722.1_67-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350471 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2735070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2937158 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253092 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2937158 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2735070 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline