Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01608460_gH
          See other OLFR293 GE Assays ›
          Gene Symbol:
          Olfr293
          Gene Name:
          olfactory receptor 293
          Gene Aliases:
          MOR221-3
          Chromosome Location:
          Chr.7: 86663664 - 86664674 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01608460_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr293
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011752.1 NP_001011752.1 1 538 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr293

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109471 NM_001011752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109471 NM_001011752.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25295387 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06380730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06380721 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06380732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25295412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06336862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06341070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06380729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06380733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25295394 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10554732 - -
          Affymetrix Mouse Gene 1.1 ST Array 10554731 - -
          Affymetrix Mouse Clariom S Array PSR0700010776.mm.1 TC0700001245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010776.mm.1 NM_001011752.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010776.mm.1 NM_001011752.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 824932 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2889746 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2889746 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline